missing translation for 'onlineSavingsMsg'
Learn More

CRIF1 Antibody, Novus Biologicals™

Product Code. 18380720 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380720 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380720 Supplier Novus Biologicals Supplier No. NBP287212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CRIF1 Polyclonal specifically detects CRIF1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias CKBBP2, CKII beta-associating protein, CR6 interacting factor 1, CRIF1p53-responsive gene 6 protein, growth arrest and DNA damage-inducible proteins-interacting protein 1, growth arrest and DNA-damage-inducible, gamma interacting protein 1, MGC4667, MGC4758, papillomavirus L2 interacting nuclear protein 1, Papillomavirus L2-interacting nuclear protein 1, Plinp1, PLINP-1CKII beta binding protein 2, PRG6CR6-interacting factor 1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of human CRIF1
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 90480
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.