missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRHR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA585638
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: U87-MG (unboiled), U87-MG MF (unboiled), Mouse brain, Rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Corticotropin-releasing factor receptor 1 (CRFR1/CRHR1), a Corticotropin-Releasing Factor Receptor, is involved in mediating the stress response and anxiety-related behaviors. Mutations of the receptor reduce anxiety-related behaviors and may be involved in the pathogenesis of Cushing disease. Multiple isoforms of CRFR1 are produced by alternative splicing. CRFR1 has been reported to be expressed in the pituitary, adrenal gland, brain, colon, ovary, placenta, testis, white blood cells, and skin. ESTs have been isolated from human brain, eye, colon, and embryo libraries.
Specifications
| CRHR1 | |
| Polyclonal | |
| Unconjugated | |
| CRHR1 | |
| corticotrophin releasing hormone receptor 1, corticotropin releasing hormone 1, corticotropin releasing hormone receptor 1, corticotropin-releasing factor receptor 1, corticotropin-releasing factor type 1 receptor, corticotropin-releasing hormone receptor 1, CRF 1 receptor, Crf receptor 1, Crf type 1, CRF1, Crf1 receptor, CRFR, CRF-R, CRFR1, CRF-R1, CRF-R-1, CRFR-1, CRF-R1alpha, CRH receptor 1, CRH receptor subtype-1 isoform-3, Crhr, CRH-R 1, CRHR1, CRH-R1, CRH-R-1, CRHR1f, CRH-R1h, CRHR1L, seven transmembrane helix receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12921, 1394, 58959 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.37 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300, pH 7 | |
| P34998, P35347, P35353 | |
| CRHR1 | |
| The immunogen used to generate this antibody corresponds to human CRHR1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction