missing translation for 'onlineSavingsMsg'
Learn More

Cresyl Diphenyl Phosphate (Mixture of Methyl-substituted Analogues), TRC

Product Code. 30357777
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
Unit Size:
10mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357777 10 mL 10mL
30365775 5 mL 5mL
30366827 1 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357777 Supplier Toronto Research Chemicals Supplier No. TRCC78301510ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26444-49-5
Molecular Formula C19 H17 O4 P
Molecular Weight (g/mol) 340.31
Synonym Phenyl tolyl phosphate ((PhO)2(C7H7)PO) (7CI), Phenyl tolyl phosphate ((PhO)2(C7H7O)PO) (6CI), Phosphoric acid, diphenyl tolyl ester (8CI), CDP, Cresol diphenyl phosphate, Cresyl DPP, Cresyl diphenyl phosphate, Diphenyl cresol phosphate, Diphenyl cresyl phosphate, Diphenyl tolyl phosphate, Disflamoll DPK, Durad CDP, Kronitex CDP, Methylphenyl diphenyl phosphate, Monocresyl diphenyl phosphate, Phosflex 112, Phosflex CDP, Phosphoric acid, cresyl diphenyl ester, Tolyl diphenyl phosphate
IUPAC Name (3-methylphenyl) diphenyl phosphate
SMILES [*]C.[*]c1c([*])c([*])c(OP(=O)(Oc2ccccc2)Oc3ccccc3)c([*])c1[*].[*H].[*H].[*H].[*H]

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C19H17O4P/c1-16-9-8-14-19(15-16)23-24(20,21-17-10-4-2-5-11-17)22-18-12-6-3-7-13-18/h2-15H,1H3
Formula Weight 340.0864
Chemical Name or Material Cresyl Diphenyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.