missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREST Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100094
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CREST.
Synovial sarcomas occur most frequently in the extremities around large joints. More than 90% of cases have a recurrent and specific chromosomal translocation, t(X;18)(p11.2;q11.2), in which the 5-prime end of the SS18 gene (MIM 600192) is fused in-frame to the 3-prime end of the SSX1 (MIM 312820), SSX2 (MIM 300192), or SSX4 (MIM 300326) gene. The SS18L1 gene is homologous to SS18.
Specifications
| CREST | |
| Polyclonal | |
| Unconjugated | |
| SS18L1 | |
| A230053O16Rik; Calcium-responsive transactivator; calcium-responsive transcription coactivator; CREST; KIAA0693; LP2261; Ss18l1; SS18L1, nBAF chromatin remodeling complex subunit; SS18-like protein 1; synovial sarcoma translocation gene on chromosome 18-like 1; SYT homolog 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192352, 26039, 269397 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75177, Q8BW22, Q91XJ0 | |
| SS18L1 | |
| A synthesized peptide derived from human SS18L1(Accession O75177), corresponding to amino acid residues S258-S308. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction