Learn More
Skytec Crest Grip Gloves
Catalog Number 17861674
Click to view available options
Size:
7, Small
8, Medium
9, Large
10, X-Large
11, 2X-Large
Crest Grip Gloves feature an ultra lightweight liner with an innovative coating providing superior grip and abrasion resistance.
  • GeniumTM is an eco friendly liner with 1/3 of the ingredients by weight that are sustainably sourced and that requires 30% less energy to be produced
  • Crinkle effect on a nitrile foam providing oustanding grip in all conditions
  • Stretch with recovery around 2-3x greater than nylon
  • Superior abrasion resistance up to 18,000 Cycles
  • No Particle shedding
  • Super soft liner
  • Anatomically designed with moisture wicking to deliver great comfort
  • Sanitized™ antimicrobial finish provides extra freshness
  • Light heat protection up to 100°C for a short period
ANSI Abrasion Rating ANSI 3
Certifications/Compliance CE CAT II
Certifications/Compliance (Europe) EN388, EN420, EN 407
Chlorinated No
Coated Area Palm
Construction Knit
Coating Material Nitrile Foam
Color Red, Black
Exterior Finish Partially Textured
Hand Specific
Latex Free No
Material Nitrile Foam, Nylon, Spandex
Product Type Grip Glove
Size 7, Small
Glove Cuff Style Elasticated
Puncture Rating ANSI/ISEA 105-16 Level 2
Resistant To Abrasion, Punctures
Thumb Style Straight