missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red, sodium salt, pure, water soluble, indicator

Product Code. 10133680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133680 5 g Glass bottle 5g
10645012 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133680 Supplier Thermo Scientific Chemicals Supplier No. 151390050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 62625-29-0 | C21H17NaO5S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-29-0
Molecular Formula C21H17NaO5S
MDL Number MFCD00001618

Specifications

Solubility Information Solubility in water: Soluble
Infrared Spectrum Authentic
Formula Weight 404.4
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 7.0 (yellow) to pH 8.8 (purple)
Color Brown
Quantity 5 g
Physical Form Powder
Chemical Name or Material Cresol Red, sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.