missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red, pure, Indicator grade

Product Code. 10103350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103350 25 g Glass bottle 25g
10467740 5 g Glass bottle 5g
10704294 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103350 Supplier Thermo Scientific Chemicals Supplier No. 151380250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1733-12-6 | C21H18O5S | 382.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1733-12-6
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005878
InChI Key OBRMNDMBJQTZHV-UHFFFAOYSA-N
PubChem CID 73013
ChEBI CHEBI:86218
IUPAC Name 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
SMILES CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

Melting Point 290.0°C
Beilstein 19, 91
Merck Index 15, 2568
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dil. acids, dil. alkalies and ethanol,, insoluble in diethyl ether, benzene
Infrared Spectrum Authentic
Formula Weight 382.42
Grade Pure
Packaging Glass bottle
Specific Extinction 1500 min. (572nm, in 0.01N NaOH)
Visual Transition Interval from pH 7.0 (orange) to pH 8.8 (purple)
Color Brown/Red or Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Cresol Red
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.