missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red Indicator, ≥83%, Solstice

Product Code. 15613830 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15613830 25 g Glass Bottle 25g
15603830 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15613830 Supplier Honeywell-Fluka Supplier No. 3265325G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1733-12-6
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005878
InChI Key OBRMNDMBJQTZHV-UHFFFAOYSA-N
Synonym o-Cresolsulfonphthalein
PubChem CID 73013
ChEBI CHEBI:86218
IUPAC Name 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
SMILES CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

UN Number NONH for all modes of transport
Beilstein 343399
Formula Weight 382.43g/mol
Loss on Drying 0 to 5%
Packaging Glass Bottle
Density 0.998g/cm3
Color Brown/Red
pH 7.2 to 8.8 (yellow to red)
Quantity 25 g
Physical Form Powder
Chemical Name or Material Cresol Red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.