missing translation for 'onlineSavingsMsg'
Learn More

Crenolanib, MedChemExpress

Product Code. 16481727
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481727 50 mg 50mg
16418267 5 mg 5mg
16405568 10 mg 10mg
16466116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481727 Supplier Medchem Express Supplier No. HY13223
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Crenolanib is a potent and selective inhibitor of wild-type and mutant isoforms of the class III receptor tyrosine kinases FLT3 and PDGFRα/β with Kds of 0.74 nM and 2.1 nM/3.2 nM, respectively.

Crenolanib is a potent and selective inhibitor of wild-type and mutant isoforms of the class III receptor tyrosine kinases FLT3 and PDGFRα/β with Kds of 0.74 nM and 2.1 nM/3.2 nM, respectively.

Specifications

Chemical Name or Material Crenolanib
CAS 670220-88-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H29N5O2
Quantity 50 mg
Synonym CP-868596
Solubility Information DMSO : 25 mg/mL (56.36 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1CCN(C2=CC=CC3=C2N=C(N4C5=CC=C(C=C5N=C4)OCC6(COC6)C)C=C3)CC1
Molecular Weight (g/mol) 443.54
Formula Weight 443.54
Percent Purity 99.5%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.