missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12131-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a bZIP transcription factor that binds to the cAMP responsive element found in many viral and cellular promoters. It is an important component of cAMP-mediated signal transduction during the spermatogenetic cycle, as well as other complex processes. Alternative promoter and translation initiation site usage allows this gene to exert spatial and temporal specificity to cAMP responsiveness. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene, with some of them functioning as activators and some as repressors of transcription.Specifications
| CREM | |
| Polyclonal | |
| Unconjugated | |
| CREM | |
| CREM, hCREM 2, ICER, Inducible cAMP early repressor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12916, 1390, 25620 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27699, Q03060, Q03061 | |
| Crem | |
| CREM Fusion Protein Ag2775 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction