missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRELD2 Recombinant Mouse Monoclonal Antibody (K1H020_9A8)
Description
Immunogen sequence: PGNCKECISG YAREHGQCAD VDECSLAEKT CVRKNENCYN TPGSYVCVCP DGFEETEDAC VPPAEAEATE GESPTQLPSR Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 72%, Pig - 72%, Cynomolgus monkey - 94%.
CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.
Specifications
Specifications
| Antigen | CRELD2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Recombinant Monoclonal |
| Clone | K1H020_9A8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide |
| Gene | CRELD2 |
| Gene Accession No. | Q6UXH1 |
| Gene Alias | 5730592L21Rik; C85758; CREL2; CRELD2; cysteine rich with EGF like domains 2; cysteine rich with EGF-like domains 2; cysteine-rich with EGF-like domain protein 2; cysteine-rich with EGF-like domains 2; DKFZp667O055; MGC11256; Protein disulfide isomerase CRELD2; RGD1310614; UNQ185/PRO211 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?