missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRELD1 Rabbit anti-Human, Clone: 116, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90218-50UL
This item is not returnable.
View return policy
Description
CRELD1 Monoclonal specifically detects CRELD1 in Human samples. It is validated for ELISA.
Specifications
| CRELD1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 78987 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 116 | |
| ELISA 1:5000-1:10000 | |
| atrioventricular septal defect 2, AVSD2, CIRRIN, cysteine-rich with EGF-like domain protein 1, cysteine-rich with EGF-like domains 1, DKFZp566D213 | |
| Recombinant Human CRELD1 protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction