missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRELD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15597942
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15597942 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15597942 Supplier Invitrogen™ Supplier No. PA547455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Epidermal growth factor (EGF; MIM 131530)-like repeats are a class of cysteine-rich domains that mediate interactions between proteins of diverse function. EGF domains are found in proteins that are either completely secreted or have transmembrane regions that tether the protein to the cell surface. CRELD1 is the founding member of a family of matricellular proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRELD1
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Creld1
Gene Accession No. Q96HD1
Gene Alias AI843811; AVSD2; CIRRIN; CRELD1; cysteine rich with EGF like domains 1; cysteine rich with EGF-like domains 1; cysteine-rich with EGF-like domain protein 1; cysteine-rich with EGF-like domains 1; DKFZP566D213; i11E7 protein; protein disulfide isomerase CRELD1; UNQ188/PRO214
Gene Symbols Creld1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRELD1 isoform 1 Gln30-Glu362.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78987
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.