missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRELD1 Antibody (116), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90218R
This item is not returnable.
View return policy
Description
CRELD1 Monoclonal specifically detects CRELD1 in Human samples. It is validated for ELISA.
Specifications
| CRELD1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 78987 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 116 | |
| ELISA | |
| atrioventricular septal defect 2, AVSD2, CIRRIN, cysteine-rich with EGF-like domain protein 1, cysteine-rich with EGF-like domains 1, DKFZp566D213 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CRELD1 (NP_001070883.1; Met1-Glu362). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction