missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566012
This item is not returnable.
View return policy
Description
Immunogen sequence: YATLTMTLAQT NFCKKHGFDP QSPLCVHIML SGTVTKVNET EMDIAKHSLF IRHPEMKTWP SSHNWFFAKL NITNIWLLD Highest antigen sequence identity to the following orthologs - mouse 78%, rat 76%.
The adenovirus E1A protein both activates and represses gene expression to promote cellular proliferation and inhibit differentiation. CREG1 antagonizes transcriptional activation and cellular transformation by E1A. This protein shares limited sequence similarity with E1A and binds both the general transcription factor TBP and the tumor suppressor pRb in vitro.The adenovirus E1A protein both activates and represses gene expression to promote cellular proliferation and inhibit differentiation. The protein encoded by this gene antagonizes transcriptional activation and cellular transformation by E1A. This protein shares limited sequence similarity with E1A and binds both the general transcription factor TBP and the tumor suppressor pRb in vitro. This gene may contribute to the transcriptional control of cell growth and differentiation.
Specifications
| CREG1 | |
| Polyclonal | |
| Unconjugated | |
| CREG1 | |
| AA755314; cellular repressor of E1A stimulated genes 1; cellular repressor of E1A-stimulated genes 1; CREG; CREG1; Protein CREG1; UNQ727/PRO1409 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75629 | |
| CREG1 | |
| Recombinant Human CREG1. Recombinant protein control fragment (Product #RP-106696). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction