missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CREBZF Polyclonal Antibody
GREENER_CHOICE

Product Code. 12829252
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829252 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829252 Supplier Invitrogen™ Supplier No. PA521106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is K562 cell lysate. PA5-21106 can be used with blocking peptide PEP-1220.

Strongly activates transcription when bound to HCFC1. Suppresses the expression of HSV proteins in cells infected with the virus in a HCFC1-dependent manner. Also suppresses the HCFC1-dependent transcriptional activation by CREB3 and reduces the amount of CREB3 in the cell. Able to down-regulate expression of some cellular genes in CREBZF-expressing cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREBZF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CREBZF
Gene Accession No. Q91ZR3, Q9NS37
Gene Alias 1110034C16Rik; 6330417B10Rik; CREB/ATF bZIP transcription factor; Crebzf; HCF-binding transcription factor Zhangfei; host cell factor-binding transcription factor Zhangfei; LAZip; LAZipII; RGD1565584; SHP-interacting leucine zipper protein; SMILE; Tyrosine kinase-associated leucine zipper protein LAZip; tyrosine kinase-associated leucine zipper protein LAZipII; ZF; Zhangfei
Gene Symbols CREBZF
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human ZHANGFEI.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233490, 58487
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.