missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREBZF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530787
This item is not returnable.
View return policy
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (88%), Rat (90%), Rhesus Monkey (97%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Strongly activates transcription when bound to HCFC1. Suppresses the expression of HSV proteins in cells infected with the virus in a HCFC1-dependent manner. Also suppresses the HCFC1-dependent transcriptional activation by CREB3 and reduces the amount of CREB3 in the cell. Able to down-regulate expression of some cellular genes in CREBZF-expressing cells.
Specifications
| CREBZF | |
| Polyclonal | |
| Unconjugated | |
| CREBZF | |
| 1110034C16Rik; 6330417B10Rik; CREB/ATF bZIP transcription factor; Crebzf; HCF-binding transcription factor Zhangfei; host cell factor-binding transcription factor Zhangfei; LAZip; LAZipII; RGD1565584; SHP-interacting leucine zipper protein; SMILE; Tyrosine kinase-associated leucine zipper protein LAZip; tyrosine kinase-associated leucine zipper protein LAZipII; ZF; Zhangfei | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 233490, 58487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q91ZR3, Q9NS37 | |
| CREBZF | |
| Recombinant protein encompassing a sequence within the center region of human CREBZF. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction