missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREBZF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101265
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CREBZF.
Strongly activates transcription when bound to HCFC1. Suppresses the expression of HSV proteins in cells infected with the virus in a HCFC1-dependent manner. Also suppresses the HCFC1-dependent transcriptional activation by CREB3 and reduces the amount of CREB3 in the cell. Able to down-regulate expression of some cellular genes in CREBZF-expressing cells.
Specifications
| CREBZF | |
| Polyclonal | |
| Unconjugated | |
| CREBZF | |
| 1110034C16Rik; 6330417B10Rik; CREB/ATF bZIP transcription factor; Crebzf; HCF-binding transcription factor Zhangfei; host cell factor-binding transcription factor Zhangfei; LAZip; LAZipII; RGD1565584; SHP-interacting leucine zipper protein; SMILE; Tyrosine kinase-associated leucine zipper protein LAZip; tyrosine kinase-associated leucine zipper protein LAZipII; ZF; Zhangfei | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 233490, 58487 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91ZR3, Q9NS37 | |
| CREBZF | |
| A synthesized peptide derived from human CREBZF(Accession Q9NS37), corresponding to amino acid residues E94-R144. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction