missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREBRF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568552
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Acts as a negative regulator of the endoplasmic reticulum stress response or unfolded protein response (UPR). Represses the transcriptional activity of CREB3 during the UPR. Recruits CREB3 into nuclear foci.
Specifications
| CREBRF | |
| Polyclonal | |
| Unconjugated | |
| CREBRF | |
| A930001N09Rik; adult retina protein; C20H5orf41; C4H5orf41; C5orf41; CREB3 regulatory factor; CREBRF; gb:bc142922; LRF; Luman recruitment factor; luman/CREB3 recruitment factor; luman-recruiting factor; RGD1310862; UPF0474 protein C5orf41; UPF0474 protein C5orf41 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 153222 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IUR6 | |
| CREBRF | |
| synthetic peptide directed towards the middle region of human CREBRF. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction