missing translation for 'onlineSavingsMsg'
Learn More

Crebinostat, TRC

Product Code. 30363825
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363825 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363825 Supplier Toronto Research Chemicals Supplier No. TRCC78152525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Crebinostat
CAS 1092061-61-4
Molecular Formula C20H23N3O3
Quantity 25 mg
Synonym 7-(Hydroxyamino)-7-oxoheptanoic acid 2-([1,1'-biphenyl]-4-ylmethylene)hydrazide, Crebinostat
InChI Formula InChI=1S/C20H23N3O3/c24-19(9-5-2-6-10-20(25)23-26)22-21-15-16-11-13-18(14-12-16)17-7-3-1-4-8-17/h1,3-4,7-8,11-15,26H,2,5-6,9-10H2,(H,22,24)(H,23,25)/b21-15+
SMILES O=C(CCCCCC(NO)=O)N/N=C/C(C=C1)=CC=C1C2=CC=CC=C2
IUPAC Name (E)-7-(2-([1,1'-biphenyl]-4-ylmethylene)hydrazineyl)-N-hydroxy-7-oxoheptanamide
Molecular Weight (g/mol) 353.41
Formula Weight 353.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.