missing translation for 'onlineSavingsMsg'
Learn More

CREB5 Antibody (PCRP-CREB5-1G8), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18352762
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352762 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352762 Supplier Bio-Techne Supplier No. NBP314073AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CREB5 Monoclonal antibody specifically detects CREB5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB5
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-CREB5-1G8
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias cAMP response element binding protein CRE-Bpa, cAMP response element-binding protein CRE-BPa, cAMP responsive element binding protein 5, cAMP responsive element binding protein 5 isoform alpha, cAMP-responsive element-binding protein 5, CREB-5, CREBPA, CRE-BPA, cyclic AMP-responsive element-binding protein 5, H_GS165L15.1
Host Species Mouse
Immunogen Recombinant full-length human CREB5 protein (Uniprot: Q02930)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9586
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.