Learn More
Invitrogen™ CREB3L4 Polyclonal Antibody, Biotin
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA5142000
Product Code. 17896972
  • £406.00 / 100µL
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_570968.1; NP_001242909.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CREB3L4
Polyclonal
Biotin
CREB3L4
1700012K17Rik; 5330432F22Rik; acre1; AIBZIP; Androgen-induced basic leucine zipper protein; androgen-induced bZIP protein; ATCE1; Attaching to CRE-like 1; AV040530; AV258827; cAMP responsive element binding protein 1; cAMP responsive element binding protein 3 like 4; cAMP responsive element binding protein 3-like 4; cAMP-responsive element-binding protein 3-like protein 4; cAMP-responsive element-binding protein 4; CREB3; Creb3l4; CREB4; CREB-4; Cyclic AMP-responsive element-binding protein 3-like protein 4; cyclic AMP-responsive element-binding protein 4; hJAL; JAL; mJAL; OTTHUMP00000035266; Processed cyclic AMP-responsive element-binding protein 3-like protein 4; RP11-422P24.8; Tisp40; transcript induced in spermiogenesis protein 40
Goat
Ammonium Sulfate Precipitation
RUO
148327
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q8TEY5
CREB3L4
Peptide with sequence CPSGRIRSVLHADEM (aa 382-395).