missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREB3L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540951
This item is not returnable.
View return policy
Description
Peptide sequence: HSYSLCEEPR AQSPFTHITS DSFNDDEVES EKWYLSTDF Sequence homology: Cow: 93%; Dog: 85%; Guinea Pig: 87%; Horse: 87%; Human: 100%; Mouse: 86%; Rabbit: 87%; Rat: 87%.
CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).
Specifications
| CREB3L2 | |
| Polyclonal | |
| Unconjugated | |
| Creb3l2 | |
| basic transcription factor 2; BBF2 human homolog on chromosome 7; BBF2H7; B-ZIB transcription factor; C530025K05Rik; cAMP responsive element binding protein 3 like 2; cAMP responsive element binding protein 3-like 2; cAMP-responsive element-binding protein 3-like protein 2; Creb3l2; Cyclic AMP-responsive element-binding protein 3-like protein 2; FUS/BBF2H7 protein; Processed cyclic AMP-responsive element-binding protein 3-like protein 2; Ra69; SCI-related protein Ra69; SCIRR69; TCAG_1951439 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 64764 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q70SY1 | |
| Creb3l2 | |
| Synthetic peptide directed towards the N-terminal of human CREB3L2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction