missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREB3L2 Polyclonal Antibody

Description
Immunogen sequence: MEVLESGEQGV LQWDRKLSEL SEPGDGEALM YHTHFSELLD EFSQNVLGQL LNDPFLSEKS VSMEVEPSPT SPAPLIQAEH SYSLCEEPRA QSPFTHITTS DSFND Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).
Specifications
Specifications
| Antigen | CREB3L2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Creb3l2 |
| Gene Accession No. | Q70SY1 |
| Gene Alias | basic transcription factor 2; BBF2 human homolog on chromosome 7; BBF2H7; B-ZIB transcription factor; C530025K05Rik; cAMP responsive element binding protein 3 like 2; cAMP responsive element binding protein 3-like 2; cAMP-responsive element-binding protein 3-like protein 2; Creb3l2; Cyclic AMP-responsive element-binding protein 3-like protein 2; FUS/BBF2H7 protein; Processed cyclic AMP-responsive element-binding protein 3-like protein 2; Ra69; SCI-related protein Ra69; SCIRR69; TCAG_1951439 |
| Gene Symbols | Creb3l2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?