missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB3L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | CREB3L2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18225295
|
Novus Biologicals
NBP2-58761 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694359
|
Novus Biologicals
NBP2-58761-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CREB3L2 Polyclonal specifically detects CREB3L2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| CREB3L2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 64764 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MEVLESGEQGVLQWDRKLSELSEPGDGEALMYHTHFSELLDEFSQNVLGQLLNDPFLSEKSVSMEVEPSPTSPAPLIQAEHSYSLCEEPRAQSPFTHITTSDSFND | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| basic transcription factor 2, BBF2 human homolog on chromosome 7, BBF2H7TCAG_1951439, B-ZIB transcription factor, cAMP responsive element binding protein 3-like 2, cAMP-responsive element-binding protein 3-like protein 2, cyclic AMP-responsive element-binding protein 3-like protein 2, FUS/BBF2H7 protein, MGC131709, MGC71006 | |
| CREB3L2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title