missing translation for 'onlineSavingsMsg'
Learn More

CREB3L1/OASIS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen CREB3L1/OASIS
Concentration 0.28 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16858173
View Documents
Proteintech
11235-2-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16848173
View Documents
Proteintech
11235-2-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Transcription factor that acts during endoplasmic reticulum stress by activating unfolded protein response target genes. Specifically involved in ER-stress response in astrocytes in the central nervous system (By similarity). May play a role in gliosis. In vitro, binds to box-B element, cAMP response element (CRE) and CRE-like sequences, and activates transcription through box-B element but not through CRE.
Specifications

Specifications

CREB3L1/OASIS
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
Rabbit
Mouse, Human, Rat
Q96BA8, Q9Z125
26427, 362165, 90993
CREB3L1,OASIS Fusion Protein Ag1757
Primary
-20°C
CREB3L1
0.28 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CREB3L1, OASIS
Creb3l1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.