missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB Antibody (5G3), Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £416.00
Specifications
| Antigen | CREB |
|---|---|
| Clone | 5G3 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18183579
|
Novus Biologicals
NBP2-37610 |
0.1 mL |
£416.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696505
|
Novus Biologicals
NBP2-37610-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CREB Monoclonal specifically detects CREB in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CREB | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Primate, Monkey | |
| active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein | |
| CREB1 | |
| IgG1 | |
| Unpurified |
| 5G3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, Transcription Factors and Regulators | |
| Ascites with 0.03% Sodium Azide | |
| 1385 | |
| Purified recombinant fragment of human CREB1 expressed in E. Coli. | |
| Primary | |
| 43 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title