missing translation for 'onlineSavingsMsg'
Learn More

Creatinine, TRC

Product Code. 30007162
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007162 1 g 1g
30003534 50 mg 50mg
30003816 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007162 Supplier Toronto Research Chemicals Supplier No. TRCC7815001G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4 H7 N3 O
Molecular Weight (g/mol) 113.118
Synonym 4H-Imidazol-4-one, 2-amino-1,5-dihydro-1-methyl-, Creatinine (8CI), 1-Methylglycocyamidine, 1-Methylhydantoin-2-imide, 2-Amino-1-methyl-1,5-dihydroimidazol-4-one, 2-Amino-1-methylimidazolin-4-one, NSC 13123, TEGO Cosmo C 250
IUPAC Name 2-amino-3-methyl-4H-imidazol-5-one
SMILES CN1CC(=O)N=C1N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)
Formula Weight 113.059
Percent Purity >95%
Chemical Name or Material Creatinine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.