missing translation for 'onlineSavingsMsg'
Learn More

Creatinine, MedChemExpress

Product Code. 16482199
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482199 1 g 1g
16410968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482199 Supplier Medchem Express Supplier No. HYB0504
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Creatinine(NSC13123) is a break-down product of creatine phosphate in muscle, and is usually produced at a fairly constant rate by the body.

Creatinine(NSC13123) is a break-down product of creatine phosphate in muscle, and is usually produced at a fairly constant rate by the body.

Specifications

Chemical Name or Material Creatinine
Physical Form Solid
CAS 60-27-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C4H7N3O
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym NSC13123
Solubility Information H2O : 33.33 mg/mL (294.64 mM; Need ultrasonic) ∣DMSO : 2.5 mg/mL (22.10 mM; Need ultrasonic)
SMILES O=C1N=C(N)N(C)C1
Molecular Weight (g/mol) 113.12
Formula Weight 113.12
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.