missing translation for 'onlineSavingsMsg'
Learn More

Creatine phosphate disodium salt tetrahydrate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022535
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
71519-72-7
C4H8N3Na2O5P·4H2O
MFCD00150192
This item is not returnable. View return policy
71519-72-7
C4H8N3Na2O5P·4H2O
MFCD00150192

Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

Solubility
Soluble in water.0.1 g/mL.

Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
TRUSTED_SUSTAINABILITY
Assay Percent Range ≥98%
Quantity 1 g
Solubility Information Soluble in water.0.1g/mL.
Formula Weight 327.14 (255.07 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Creatine Phosphate Disodium Salt Tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'