missing translation for 'onlineSavingsMsg'
Learn More

Creatine, MedChemExpress

Product Code. 16452678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452678 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452678 Supplier Medchem Express Supplier No. HYW010388
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Creatine, an endogenous amino acid derivative, plays an important role in cellular energy, especially in muscle and brain.

Creatine, an endogenous amino acid derivative, plays an important role in cellular energy, especially in muscle and brain.

Specifications

Chemical Name or Material Creatine
CAS 57-00-1
Color White
Purity Grade Notes Research
Molecular Formula C4H9N3O2
Quantity 5 g
Solubility Information H2O : 6.25 mg/mL (47.66 mM; Need ultrasonic)
SMILES O=C(O)CN(C)C(N)=N
Molecular Weight (g/mol) 131.14
Formula Weight 131.14
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.