missing translation for 'onlineSavingsMsg'
Learn More

Creatine Kinase, Muscle/CKMM Mouse anti-Human, Clone: CKMM/3341, Novus Biologicals™

Mouse Monoclonal Antibody

£377.00

Specifications

Antigen Creatine Kinase, Muscle/CKMM
Clone CKMM/3341
Concentration 0.2 mg/mL
Dilution ELISA, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18643649
View Documents
Novus Biologicals
NBP3-07560-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Creatine Kinase, Muscle/CKMM Monoclonal specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for ELISA, Protein Array.
Specifications

Specifications

Creatine Kinase, Muscle/CKMM
0.2 mg/mL
ELISA, Peptide Array
Unconjugated
Mouse
Human
CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK
Recombinant full-length Creatine Kinase, Muscle/CKMM protein
Primary
Store at 4°C.
CKMM/3341
ELISA, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
1158
IgG2a κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.