missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CKMT1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560387
This item is not returnable.
View return policy
Description
Immunogen sequence: YLIDCERRLE RGQDIRIPTP VIHTKH Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 81%.
Mitochondrial creatine kinase (MtCK) is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Many malignant cancers with poor prognosis have shown overexpression of ubiquitous mitochondrial creatine kinase, this may be related to high energy turnover and failure to eliminate cancer cells via apoptosis. Ubiquitous mitochondrial creatine kinase has 80% homology with the coding exons of sarcomeric mitochondrial creatine kinase.
Specifications
| CKMT1A | |
| Polyclonal | |
| Unconjugated | |
| CKMT1A | |
| Acidic-type mitochondrial creatine kinase; CKMT; Ckmt1; CKMT1A; Ckmt1b; Creatine kinase U-type, mitochondrial; creatine kinase, mitochondrial 1 (ubiquitous); creatine kinase, mitochondrial 1, ubiquitous; creatine kinase, mitochondrial 1A; creatine kinase, mitochondrial 1B; mia-CK; mi-CK; Mt-CK; ScCKmit; UbCKmit; Ubiquitous mitochondrial creatine kinase; U-MtCK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 548596 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12532 | |
| CKMT1A | |
| Recombinant protein corresponding to Human CKMT1A. Recombinant protein control fragment (Product #RP-99584). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction