missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine kinase MT 1A, p Tyr153 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-60776
This item is not returnable.
View return policy
Description
Creatine kinase MT 1A Polyclonal specifically detects Creatine kinase MT 1A in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot.
Specifications
| Creatine kinase MT 1A (p Tyr153) | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| Acidic-Type Mitochondrial Creatine Kinase, CKMT, CKMT1, CKMT1A, Creatine Kinase U-Type, Mitochondrial, Creatine Kinase, Mitochondrial 1 (Ubiquitous), Creatine Kinase, Mitochondrial 1A, EC 2.7.3.2, Mia-CK, Ubiquitous Mitochondrial Creatine Kinase, U-MtCK | |
| Rabbit | |
| 75 kDa | |
| 100 μL | |
| 548596 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
| CKMT1A | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr153 conjugated to KLH. Accession # P12532 | |
| Antigen Affinity-purified | |
| Primary | |
| Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction