missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Creatine Kinase MM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586374
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifications
| Creatine Kinase MM | |
| Polyclonal | |
| Unconjugated | |
| CKM | |
| CKM; CKMM; CPK-M; Creatine kinase M chain; creatine kinase M-type; creatine kinase, M-type; creatine kinase, muscle; creatine phosphokinase M-type; M creatine phosphokinase; MCK; M-CK; muscle creatine kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1158, 12715, 24265 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00564, P06732, P07310 | |
| CKM | |
| Synthetic peptide, corresponding to the N-terminus of Human Creatine Kinase M. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction