Learn More
Invitrogen™ Creatine Kinase MB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113731
Description
Immunogen sequence: GVDTAAVGSV FDVSNADRLG SSEVEQVQLV VDGVKLMVEM EKKLEKGQSI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifications
| Creatine Kinase MB | |
| Polyclonal | |
| Unconjugated | |
| Ckb | |
| Bck; B-CK; Brain creatine kinase; CK MB; Ck3; Ck-3; CKB; Ckbb; Ckbr; Ckm; CKMM; CPK-B; CPK-M; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, M-type; creatine kinase, muscle; creatine kinase-B; creatine kinase-M; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29; MCK; M-CK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1152, 1158, 12709, 12715 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P06732, P07310, P12277, Q04447 | |
| Ckb, CKM | |
| Synthetic peptide directed towards the middle region of human CKM. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.