missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase MB (CK-MB), Mouse anti-Human, Clone: 1F1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB0344-M02.100ug
This item is not returnable.
View return policy
Specifications
| Creatine Kinase MB (CK-MB) | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 | |
| Native purified human Creatine Kinase MB. | |
| RUO | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 1F1 | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction