missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase MB Antibody (1F2/1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62195DL594
This item is not returnable.
View return policy
Description
Creatine Kinase MB Monoclonal antibody specifically detects Creatine Kinase MB in Human samples. It is validated for ELISA
Specifications
| Creatine Kinase MB | |
| Monoclonal | |
| DyLight 594 | |
| Creatine Kinase Isoenzyme | |
| Highly purified human CKMB. | |
| 0.1 mL | |
| metabolism | |
| 1152 | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA | |
| 1F2/1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction