Learn More
Invitrogen™ Creatine Kinase BB Monoclonal Antibody (CK-CYK/21E10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533339
Description
Product is supplied at 1 mL (100 μL/mL). Application notes: Western blot - Under reducing conditions, antibody detected a protein with a band size of 43-46 kDa. IHC-F- tissue was fixed for 2 hours in 2% paraformaldehyde and 0.1 % glutaraldehyde in 0.1 M PB. The tissue was stored until use in 1 % paraformaldehyde. The tissue was blocked with 1% normal swine serum. IHC-P - antigen retrieval: 0.5 % pepsin in 0.01 N HCl for 15 min at 37 °C; endogenous peroxidise blocking by using 3 % H2O2 in PBS for 30 min; blocking with 1% normal swine serum for 30 min.; Purkinje cells serve as positive control and connective tissue as negative control. Positive control: Brain lysate, Human Purknije cells; Negative control: Human fibroblasts.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifications
| Creatine Kinase BB | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| P00567, P07335, P12277, Q04447 | |
| CKB | |
| Synthetic 17-mer peptide corresponding to the N-terminal sequence of human creatine kinase brain-type (SNSHNALKLRFPAEDEF). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CK-CYK/21E10 | |
| Unconjugated | |
| CKB | |
| Bck; B-CK; brain creatine kinase; Ck3; Ck-3; CKB; Ckbb; Ckbr; CPK-B; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100009085, 1152, 12709, 24264 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.