missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Creatine Kinase BB Monoclonal Antibody (CK-BYK/21E10), Biotin
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549030
This item is not returnable.
View return policy
Description
Creatine Kinase BB Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifications
| Creatine Kinase BB | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| P12277 | |
| CKB | |
| Synthetic 17-mer peptide corresponding to the N-terminal sequence of human creatine kinase brain-type (SNSHNALKLRFPAEDEF) Aliases Creatine kinase B chain, B-CK. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CK-BYK/21E10 | |
| Biotin | |
| CKB | |
| Bck; B-CK; brain creatine kinase; Ck3; Ck-3; CKB; Ckbb; Ckbr; CPK-B; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29 | |
| Mouse | |
| Protein G | |
| RUO | |
| 1152 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction