missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Creatine Kinase BB Polyclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Host Species | Rabbit |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?