missing translation for 'onlineSavingsMsg'
Learn More

Creatine Kinase BB Antibody (CPTC-CKB-2) - Azide and BSA Free, Novus Biologicals™

Product Code. 18360773 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360773 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360773 Supplier Novus Biologicals Supplier No. NBP308866

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Creatine Kinase BB Monoclonal specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Creatine Kinase BB
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-CKB-2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias Creatine Kinase Isoenzyme
Host Species Mouse
Immunogen Recombinant human full-length Creatine Kinase BB protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1152
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.