missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase BB Antibody (CKBB/6565) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21074JF585
This item is not returnable.
View return policy
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Creatine Kinase BB | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Creatine Kinase Isoenzyme | |
| Recombinant human full-length Creatine Kinase BB protein | |
| 0.1 mL | |
| Cancer | |
| 1152 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| CKBB/6565 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction