missing translation for 'onlineSavingsMsg'
Learn More

Creatine Kinase BB Antibody (CKB/1268), Novus Biologicals™

Mouse Monoclonal Antibody

£210.00 - £373.00

Specifications

Antigen Creatine Kinase BB
Clone CKB/1268
Concentration 0.2 mg/mL
Applications ELISA, Protein Array
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18665416
View Documents
Novus Biologicals
NBP2-75762-20ug
20 μg
£210.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18655747
View Documents
Novus Biologicals
NBP2-75762-100ug
100 μg
£373.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

Creatine Kinase BB
0.2 mg/mL
Monoclonal
Purified
RUO
Human
P12277, P12277, P12277, P12277
1152
IgG1 κ
Protein A or G purified
CKB/1268
ELISA, Protein Array
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
Creatine Kinase Isoenzyme
Recombinant full-length human Creatine Kinase BB protein (Uniprot: P12277)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.