missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase B Mouse anti-Human, Clone: 8.00E+04, Novus Biologicals™
Mouse Monoclonal Antibody
£241.00 - £378.00
Specifications
| Antigen | Creatine Kinase B |
|---|---|
| Clone | 80000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1063, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288742
|
Novus Biologicals
NBP2-59462-50UL |
50 μL |
£241.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246445
|
Novus Biologicals
NBP2-59462-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Creatine Kinase BB Monoclonal specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Creatine Kinase B | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 1152 | |
| Recombinant human CKB (1-381aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 80000 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1063, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-CK, Creatine kinase B chain, creatine kinase, brain, creatine kinase-B, EC 2.7.3, EC 2.7.3.2 | |
| CKB | |
| IgG2a κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title