missing translation for 'onlineSavingsMsg'
Learn More

Cre Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £399.00

Specifications

Antigen Cre
Dilution Western Blot 1:1000 - 1:2000. Use reported in scientific literature (PMID 33784255), Immunohistochemistry 1:10 - 1:5000, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:10 - 1:5000, SDS-Page reported in scientific literature (PMID 27432829), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25429143)
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18575572
View Documents
Novus Biologicals (Bio-Techne)
NB100-56134
0.05 mL
£399.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18584193
View Documents
Novus Biologicals
NB100-56134-0.025ml
0.025 mL
£178.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cre Polyclonal antibody specifically detects Cre in Mouse, Cre/lox System samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page, Immunohistochemistry Free-Floating
Specifications

Specifications

Cre
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Rabbit
Mouse, Cre/lox System
Cyclization recombinase
IgG
Unpurified
Western Blot 1:1000 - 1:2000. Use reported in scientific literature (PMID 33784255), Immunohistochemistry 1:10 - 1:5000, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:10 - 1:5000, SDS-Page reported in scientific literature (PMID 27432829), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25429143)
Polyclonal
Purified
RUO
50 uL neat whole antisera
A synthethic peptide corresponding to a portion of amino acids 1-50 of the bacteriophage protein CRE was used as immunogen, GenBank no. AAQ14086.1.
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.