missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRCP Recombinant Rabbit Monoclonal Antibody (K01_2Q42)

Product Code. 30827805
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827805 100 μL
30826618 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827805 Supplier Invitrogen™ Supplier No. MA564267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CRCP Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRCP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2Q42
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene CRCP
Gene Accession No. O35427, O75575, Q8VHM6
Gene Alias AL022669; calcitonin gene-related peptide-receptor component protein; CGRP receptor component; CGRPRCP; CGRP-RCP; CGRP-receptor component protein; Crcp; DNA-directed RNA polymerase III subunit RPC9; RCP; RCP9; RNA polymerase III subunit C9
Gene Symbols CRCP
Host Species Rabbit
Immunogen A synthetic peptide of human CRCP.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114205, 12909, 27297
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.