missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRBN Recombinant Rabbit Monoclonal Antibody (PSH0-69), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA549359
This item is not returnable.
View return policy
Description
Sequence Similarities: 95% Mouse/Rat. Tissue Specificity: Widely expressed. Highly expressed in brain. Positive Control: HEK-293 cell lysate, A375 cell lysate, A549 cell lysate, HeLa cell lysate, SH-SY5Y cell lysate, NIH/3T3 cell lysate, MCF7 cell lysate, mouse brain tissue lysate, rat testis tissue lysate, human testis tissue, human brain tissue. Subcellular Location: Cytoplasm, Nucleus, Membrane. Predicted band size: 51 kDa.
CRBN, a member of the Lon protease protein family, plays a role in brain development. It is a component of the DCX (DDB1-CUL4-X-box) E3 protein ligase complex, a complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins and is required for limb outgrowth and expression of the fibroblast growth factor FGF8. CRBN is thought to regulate the assembly and neuronal surface expression of large-conductance calcium-activated potassium channels in brain regions involved in memory and learning via its interaction with KCNT1. It is widely expressed and highly expressed in brain.Specifications
CRBN | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q56AP7, Q8C7D2, Q96SW2 | |
CRBN | |
Recombinant protein within human crbn aa 1-442 / 442. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
PSH0-69 | |
Unconjugated | |
CRBN | |
2610203G15Rik; 2900045O07Rik; AD-006; AF229032; AW108261; cereblon; CRBN; DKFZp781K0715; MGC27358; MRT2; MRT2A; novel lethal; piL; protein cereblon; Protein PiL; protein x 0001 | |
Rabbit | |
Protein A | |
RUO | |
297498, 51185, 58799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |