missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CRBN Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Specifications
Specifications
| Accession Number | NP_057386 |
| Concentration | 0.2 mg/ml |
| For Use With (Application) | Block/Neutralize |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin |
| Gene ID (Entrez) | 51185 |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | cereblon, DKFZp781K0715, MGC27358, MRT2A, non-syndromic, autosomal recessive, 2A, protein cereblon, protein x 0001 |
| Gene Symbol | CRBN |
| Research Category | Neurodegeneration, Neuroscience |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction