missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560462
This item is not returnable.
View return policy
Description
Immunogen sequence: RWDDGLRHLV MLSFGPDQLQ DLGQHVHVGG RLLAADSQPW GGPFRGCLQD LRLDGCHLPF FPLPLDNSSQ PSELGGRQSW NLT Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.
CRB2 (Crumbs homolog 2), like its homologs CRB1 and CRB3, is similar to the Drosophila crumbs protein and is expressed in retina, brain and kidney. Along with other proteins, the Crumbs proteins form a complex that help set up cell polarity in developing neuroepithelial cells. At the onset of neural specification, embryonic stem cells (ESCs) upregulate CRB2, which then localizes apically in neural rosettes. Gain- and loss-of-function studies of CRB2 have shown that CRB2 is essential for the stabilization of other polarity proteins. Unlike CRB1, mutations in CRB2 do not appear to play a role in retinitis pigmentosa or in Leber congenital amaurosis.
Specifications
| CRB2 | |
| Polyclonal | |
| Unconjugated | |
| CRB2 | |
| 5930402A21; BC043114; Crb2; crumbs 2; crumbs 2, cell polarity complex component; crumbs family member 2; crumbs homolog 2; crumbs2; crumbs-like protein 2; FSGS9; Protein crumbs homolog 2; RGD1309368; VMCKD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 286204 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5IJ48 | |
| CRB2 | |
| Recombinant protein corresponding to Human CRB2. Recombinant protein control fragment (Product #RP-100819). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction